Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Fluorescein, disodium salt dihydrate, 90%, pure

Catalog No. AC410641000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

Catalog No. AC410641000

Supplier: Thermo Scientific Chemicals 410641000

Only null left
Add to Cart

CAS: 1264198-62-0 | C20H10Na2O5·2H2O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1264198-62-0
Molecular Formula C20H10Na2O5·2H2O
MDL Number MFCD00150814
Synonym Acid Yellow 73 dihydrate, Fluorescein, water soluble dihydrate, Uranine dihydrate, C.I. 45350 dihydrate

Specifications

Assay Percent Range 90%
Beilstein 19, 222
Merck Index 14, 4159
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 412.3
Percent Purity 90%
Grade Pure
Packaging Glass bottle
UV_Vis 88% min. (on dry substance)
Color Brown to Orange/Red or Brown/Green
Quantity 100 g
Physical Form Powder
Chemical Name or Material Fluorescein, disodium salt dihydrate, 90%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-253-0
RTECSNumber LM5425000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.