Learn More
Chem Service Inc Fluopyram, 658066-35-4, MFCD17166960, 50 mg
Supplier: Chem Service Inc N1398850MG
Molecular Formula: C16 H11 Cl F6 N2 O, Molecular Weight: 396.717, Classification: FUNGICIDE, Synonym: N-[2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]ethyl]-2-(trifluoromethyl)benzamide, N-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]ethyl}-α,α,α-trifluoro-o-toluamide. Fluopyram is a new pyridinylethylbenzamide fungicide. It is highly efficient in controlling a wide variety of pathogens such as Botrytis spp., Sclerotinia spp., and Monilia spp., and also against powdery mildews and some leaf spot diseases in a broad range of crops.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.