Learn More
Perkin Elmer US LLC FlowMark Electronic Flowmeter
Supplier: Perkin Elmer US LLC N9307086
PerkinElmers FlowMark flowmeter is specifically designed for use with gas chromatography (GC) instruments. The probe is applied directly to the gas flow stream and the measured flow rate is presented on the LCD screen in units of mL/min. This unit provides continuous real-time measurements of gas streams ranging from 0.50 mL/min to 500 mL/min. Because the technology uses volumetric flow measurement, the unit is compatible with all laboratory gases. The flowmeter is designed to measure clean, dry, non-corrosive gases. Safety features include NIST traceable calibration, over-range indicator, auto shut-off, and explosion-proof rating for flammable and explosive gas atmospheres. A storage case is included and a recalibration service is available (N9307085). Technical Specifications: Accuracy: +/-2% of flow or +/-0.2 mL/min, whichever is greater Warranty: 1 year Certifications: CE, Ex Compliance: WEEE, RoHS Battery Requirements: 2-AA Batteries Data Output: USB Port
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.