Learn More
FITC (Fluorescein isothiocyanate) Mouse anti-Chemical, FITC, Clone: F4/1, Exalpha

Description
Specifications
Specifications
| Antigen | FITC (Fluorescein isothiocyanate) |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | F4/1 |
| Concentration | 0.5-2 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 0.08% sodium azide; pH |
| Gene Alias | Fluorescein |
| Host Species | Mouse |
| Immunogen | F4/1 is a hybridoma formed by fusing SP-2/0 Agl4 cells with spleen cells of a BALB/c mouse that had been immunised with FITC-KLH. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.