Learn More
Description
Includes
Clamp
LED magnifier lamp
- Useful in inspection, electronics, quality control, sample examination, and biological analysis
- Brilliant LED light closely resembles daylight for optimal color detection and clarity
- Large viewing area (4 x 2½ inches) is easy to read and arm extension is 31 inches
- Clamps vertically or horizontally to top or side of any edge
Specifications
Specifications
| Product Type | Magnifier Lamp, LED |
| Magnification | 3x |
| Color | White |
| For Use With (Application) | Inspection, electronics, quality control, sample examination, and biological analysis |
| Includes | Clamp |
| Lighting | LED |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
