Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Check Valves

Catalog No. p-200352 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With:
Tubing ID 1/4 to 3/8 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
15-315-33A Tubing ID 1/4 to 3/8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-315-33A
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Polypropylene valves prevent reverse flow in pressure or vacuum system.

 

  • Tapered, serrated ends fit several tube sizes
  • Withstand temperatures to 100°C
  • Nominal OD: 8/9/10 mm 

 

Specifications

Autoclavable Autoclavable
Material Polypropylene
For Use With Tubing ID 1/4 to 3/8 in.
Color Transparent
Product Type Check Valve
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.