Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ Decimal Stopwatch

Catalog No. p-4700005 Shop All Fisherbrand Products
Click to view available options
Includes:
Stopwatch, lanyard, 3V battery
Stopwatch, lanyard, battery
Battery Type:
Lithium Cell, 3V (CR2032)
Silver Oxide battery, 4-yr.

Catalog No. 14-649

Includes

Stopwatch, lanyard

Precise, Traceable timing for enzyme tests, viscosity measurements, animal studies, and all timed lab tests. Fisherbrand Traceable Decimal Stopwatch is tough with water-resistant ABS plastic case which seals out moisture, dust, and fumes encountered in harsh lab, plant, or field environments.

Made for scientists in industrial, educational, hospital, and governmental labs who insist on the finest instrumentation. Engineered for ease of operation, accuracy, and reliability.

  • Quartz-crystal design is accurate to 0.0005% — over 100 times more accurate than other stopwatches
  • Decimal stopwatch's four timing formats/resolution are: hr./min./sec.; 1/100 sec.; 1/1000 min; 1/100,000 hr.
  • Maximum readouts are 19.999999 hours, 1199.999 minutes, 71999.99 seconds, or 19 hr./59 min./59 sec./99 hundredths
  • Stopwatch functions include single-action, time-out, interval-split, cumulative split, and continuous roll-over timing
  • Triple display shows cumulative splits, interval splits, and running time
  • 500 recallable memories display the lap, split and running time
  • Memory feature also displays the fastest, slowest, and average lap time
  • Unit displays time of day, date, day of the week and features a time-of-day alarm
  • Additional features include countdown alarm, countdown then countup alarm, countdown repeat, pacer, and stroke functions
  • Timing range is 19 hours, 59 minutes, 59 seconds, and 99 hundredths
  • Accuracy: 0.0005%
  • Size: 6.4 x 7.6 x 1.9 cm (2.5 x 3 x 0.75 in.)
  • Weight: 2.5 oz.

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.

Specifications

Product Type Digital Stopwatch
Time Range 20 hr.
Alarms Audible
Accuracy 5e-06
Resolution 0.01 sec.
Display LCD
Battery Type Silver Oxide battery, 4-yr.
Dimensions (L x W x H) 2.5 x 3 x 0.75 in. (6.35 x 7.62 x 1.9 cm)
Length (English) 2.5 in.
Length (Metric) 6.35 cm
Width (English) 3 in.
Width (Metric) 7.62 cm
Height (English) 0.75 in.
Height (English) Display 0.75 in.
Height (Metric) 1.9 cm
Height (Metric) Display 1.9 cm
Weight (English) 2.5 oz.
Weight (Metric) 0.07 kg
Includes Stopwatch, lanyard, battery
For Use With (Application) Enzyme tests, viscosity measurements, animal studies and all timed lab tests
Certifications/Compliance ISO 17025, A2LA, NIST
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.