Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ Signature Mono Test Kit

Catalog No. 23200275 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 Tests
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Catalog No. 23-200-275

Edge

Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Includes

Test sticks, reagents, controls, workstation, package insert, capillary tubes, transfer pipettes and test tubes

Sure-Vue™ Signature Mono Test Kit includes a rapid, lateral flow test, waived for whole blood.

  • For qualitative detection of infectious mononucleosis heterophilic antibodies in serum, plasma or whole blood
  • CLIA Complexity: Whole blood-Waived; Serum/Plasma-Moderate
  • CPT Code: 86308
  • Sensitivity: 100%
  • Specificity: 96.0%
  • Test Time: five minutes
  • Internal procedural controls and positive and negative external controls included
  • Sample Types: Serum, plasma, whole blood
TRUSTED_SUSTAINABILITY

Specifications

Accuracy >99% Sensitivity
Content And Storage Room Temperature (15°C to 30°C, 59°F to 86°F)
Control Sets Included
CPT Code 86308QW (Whole Blood), 86308 (Serum or Plasma)
Clia Complexity Waived (Whole Blood), Moderate (Serum and Plasma)
Detectable Analytes Infectious Mononucleosis Heterophil Antibodies
Format Immunochromatographic Dipstick
Sample Type Serum, Plasma, Whole Blood
Sensitivity >99%
Type Rapid Test
Includes 25 test sticks, 25 test tubes, 25 transfer pipettes, 25 capillary pipettes, 1 diluent, 1 Mono positive control, 1 Mono negative control, 1 workstation, 1 instructions for use (IFU)
Packaging Type Test Kit
Quantity 25 Tests
Specificity 96%
Test Time 5 min.
Analysis Time 5 min.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.