Learn More
Braintree Scientific Fish Fin Ear Punch Pincher, 2.75mm punch
SDP
Supplier:  Braintree Scientific FFPAQUA7075
Encompass
Fish Fin Punch Pincher - These pinchers were developed after having several testimonies from customers with sore wrists and thumbs from using small ID punchers. The Fish Fin Punch Pincher relieves stress on wrist during ID marking as it requires a very gentle pressure to cut a hole. Stainless steel puncher has a sloped edge which provides a cut rather than a punch, which minimizes animal discomfort. Autoclavable, but should be lubricated afterwards for long and trouble free usage. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4840