Learn More
Omega Scientific Inc Fetal Bovine Serum, Usda Approved, 500 Ml
Supplier: Omega Scientific Inc FB01
FB-01: Fetal Bovine Serum, USDA Approved, 500 mL- Virus Screened- Mycoplasma Tested- Triple 0.1 Micron Filtered-Low Hemoglobin lots (<15mg/dL) may be available.-Low Endotoxin lots (<1.0EU/mL) may be available.Ships on dry ice