Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ RNase H (5 U/μL)
GSA_VA

Catalog No. feren0202
Click to view available options
Quantity:
100 units
500 units

Catalog No. FEREN0202

Supplier: Thermo Scientific™ EN0202

Only null left
Add to Cart
Add to Cart

Ribonuclease H (RNase H) is an endoribonuclease that specifically degrades the RNA strand in RNA-DNA hybrids.

 Ribonuclease H (Rnase H) specifically degrades the RNA strand in RNA-DNA hybrids. It does not hydrolyze the phosphodiester bonds within single-stranded and double-stranded DNA and RNA.

  • Does not hydrolyze the phosphodiester bonds within single-stranded and double-stranded DNA and RNA
Quality Control:
  • The absence of endo-, exodeoxyribonucleases and ribonucleases confirmed by appropriate quality tests

Source:

  • E.coli MRE-600 cells

Molecular Weight:

  • 18.4kDa monomer

Definition of Activity Unit:

  • One unit of the enzyme catalyzes the formation of 1nmol of acid soluble products in 20 min. at 37°C
  • Enzyme activity is assayed in the following mixture: 20mM Tris-HCl (pH 7.8), 40mM KCl, 8mM MgCl2, 1mM DTT, 24μM [3H]-poly(A)·poly(dT), 0.03mg/mL BSA, 4% (v/v) glycerol
Storage Buffer:
  • The enzyme is supplied in: 25mM HEPES-KOH (pH 8.0), 50mM KCl, 0.1mM EDTA, 1mM DTT, 0.1mg/mL BSA and 50% (v/v) glycerol

10X Reaction Buffer:

  • 200mM Tris-HCl (pH 7.8), 400mM KCl, 80mM MgCl2, 10mM DTT

Inhibition and Inactivation:

  • Inhibitors: metal chelators, SH-blocking reagents
  • Inactivated by heating at 65°C for 10 min.

Recommended for:

  • Removal of mRNA prior to synthesis of second strand cDNA (1)
  • RT-PCR and qRT-PCR: removal of RNA after first strand cDNA synthesis
  • Removal of the poly(A) sequences of mRNA after hybridization with oligo(dT) (2)
  • Site-specific cleavage of RNA (3)
  • Studies of in vitro polyadenylation reaction products

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Enzyme RNase
Compatible Buffer 10X Reaction Buffer
Quantity 500 units
Product Type RNase H

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.