Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneJET PCR Purification Kit
GSA_VA

Catalog No. FERK0701
Click to view available options
Quantity:
250 Preps
50 Preps

Catalog No. FERK0701

Supplier: Thermo Scientific™ K0701

Only null left
Add to Cart
Edge
Add to Cart

Thermo Scientific GeneJET PCR Purification Kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.

Thermo Scientific GeneJET PCR Purification Kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions. It effectively removes primers, dNTPs, unincorporated labeled nucleotides, enzymes, and salts from PCR and other reaction mixtures.

The kit can be used for purification of DNA fragments from 25 bp to 20 kb with recovery rates up to 100%. Each GeneJET purification column has a total binding capacity of up to 25 μg of DNA, and the entire procedure takes 5 minutes. The purified DNA can be used in common downstream applications, such as sequencing, restriction digestion, labeling, ligation, cloning, in vitro transcription, blotting or in situ hybridization.

Highlights

  • Highly efficient: 90 to 100% recoveries in the range of 100 bp to 10 kb
  • Fast: procedure takes only 5 minutes
  • Convenient: spin columns are capped and assembled with collection tubes

Applications

Fast and efficient purification of DNA fragments ideal for use in all conventional molecular biology procedures including:

  • FastDigest or conventional restriction digestion
  • Automated fluorescent or radioactive sequencing
  • PCR
  • In vitro transcription
  • In situ hybridization
  • Labeling
  • Cloning

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.
Format Spin Column
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line GeneJET
Product Type PCR Purification Kit
Quantity 50 Preps
Run Time 5 min.
Target DNA from Enzymatic Reactions (e.g. PCR)
For Use With (Application) Real Time PCR (qPCR), Cloning, DNA Sequencing
Isolation Technology Spin Column
Sample Type dsDNA
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.