Learn More
Selleck Chemical LLC Fasudil (HA-1077) HCl 500mg 105628-07-7
SDP
Supplier:  Selleck Chemical LLC S1573500MG
Encompass_Preferred
Fasudil(HA-1077), a potent and selective inhibitor of Rho kinase, displays less potent inhibiton over PKA, PKG, PKC and MLCK with Ki of 1.6, 1.6, 3.3, and 36 ?M in cell-free assays, respectively. Fasudil is also a calcium channel blocker. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-832-1