Learn More
Excelta™ Tweezer Vac Electric Vacuum System
Catalog No. 17468507 Shop All Excelta Corporation Products
Click to view available options
:
With high precision operation
Catalog No. 17-468-507
Supplier: Excelta™ PV100
With high precision operation
  • The PV-100 push button vacuum pen system pulls 6 inches of mercury and comes with 9 vacuum tips and no skid rubber feet.
  • Kit includes the following cup/probe assemblies: PVB-CB-18, PVB-CB-14, PVB-CB-332, PVB-CB-316, PVB-CB-38, PVB-CB-18, PVB-CB-14, PVB-CB-38 and PVB-CB-12.
Specifications
Product Type Vacuum System
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.