Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Eucaryotic Cell Biology Experiment Kit
SDPSCI_ED

Catalog No. S42877 Shop All EDVOTEK Products
Click to view available options
:
EdvoTek™ Eucaryotic Cell Biology Experiment Kit

Catalog No. S42877

Supplier: Edvotek™ 1001

Familiarizes students with simple, robust cell culture system.

  • Used in life science research, biotechnology laboratories
  • Provides student with skill set desired in both academic research and industry
  • Contains live materials

Time requirements:

  • Basic cell culture techniques - 30 minutes
  • Examination of Insect Cell Cultures - 30 minutes
  • Maintenance of Insect Cell Cultures - 1 hour
  • Cell Viability using Trypan Blue - 30 minutes
  • Differential staining using Giemsa stain - Overnight

Specifications

Product Type Eucaryotic Cell Biology Experiment Kit
Grade 9 to Undergraduate
For Use With (Application) Educational purposes
Includes Instructions, growth media, practice media, flasks, Giemsa stain, Trypan blue dye, sterile T25 flasks, sterile culture dishes, sterile large pipettes, small pipets, cell counting chambers, Sf9 insect cells.
Time Range 2.5 hr.
Content And Storage Refrigeration needed
Class Size 6 Lab Groups
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.