Learn More
Invitrogen™ EPR1 Polyclonal Antibody

Description
EPR1 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | EPR1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene Alias | EPR-1 |
| Host Species | Rabbit |
| Immunogen | KLH conjugated synthetic peptide between 308-337 amino acids from the C-terminal region of human EPR1. |
| Purification Method | Ammonium sulfate precipitation, Size-exclusion - Dialysis |
| Quantity | 400 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.