Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ EpiLife™ Medium, with 60 μM calcium

Click to view available options
Quantity:
500 mL
Description
A sterile, liquid medium prepared with 60μM calcium chloride for the long-term, serum-free culture of human epidermal keratinocytes and human corneal epithelial cells. This basal medium requires the addition of an appropriate growth supplement prior to use. EpiLife™ greatly extends the in vitro lifespan of human epidermal keratinocytes compared to other serum-free media systems. This basal media is animal origin free and contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. It does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. EpiLife™ medium is HEPES and bicarbonate buffered, and is designed for use in an incubator with an atmosphere of 5% CO2 and 95% air.
Specifications
Specifications
| Cell Type | Keratinocyte Cells, Epithelial Cells |
| Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
| Form | Liquid |
| Product Type | EpiLife Medium |
| Serum Level | Serum-free |
| Sterility | Sterile-filtered |
| Sterilization Method | Sterile-filtered |
| With Additives | HEPES, Phenol Red |
| Product Line | Cascade Biologics, EpiLife |
| Quantity | 500 mL |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction