Learn More
ENG Scientific Eosinophil Stain Kit
Catalog No. ES6920
Click to view available options
Quantity:
3 x 250 mL
This item is not returnable. View return policy
Catalog No. ES6920
Supplier: ENG Scientific 6920
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used in staining eosinophils from secretions of the nasal passage, sinuses, bronchi and urinary sediment. For Eosinophil Stain Kit. Contains Solution I - Eosin Y, Solution II - Buffer solution and Solution III - Methylene Blue solution.
  • Eosinophils stain with red cytoplasm and bright red granules; nucleus stains pale blue
  • Solutions made from certified dyes (when applicable)
  • Eosinophil Stain Kit
  • Includes: Contents include Eosin Y, Methanol, Buffer Salts, Methylene Blue
Specifications
Quantity 3 x 250 mL
Includes Solutions I - III
Packaging Bottles
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.