Learn More
Ansell ENCORE™ Perry Style 42™ PF Surgical Gloves
Catalog No. 19501151 Shop All Ansell Products
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
This item is not returnable. View return policy
Catalog No. 19-501-151
Supplier: Ansell 5711100PF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Latex, powder free surgical gloves feature a smooth finish and optimized thickness. Ideal for brief, low-risk sterile procedures inside or outside of the operating room.
  • Natural rubber latex
  • Made with proprietary anatomic formers for comfort and to help reduce hand fatigue
  • Polymer coated for fast and easy donning
  • Optimized glove thickness for the best balance of protection and tactile sensitivity
  • Recommended for:
    • Brief, low-risk examination procedures
    • Sterile procedures in the O.R.
    • Use as an outer glove when double gloving
    • Sterile procedures outside of the O.R.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM D3577
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
AQL 0.65
Color Natural
Exterior Finish No Texture
Laboratory/Cleanroom Use No
Material Latex
Size 5.5
Sterility Sterile
Manufacturing/QMS Audit Standards ISO 13485, ISO 9001
Model 5711100PF
Series ENCORE™ Perry Style 42™ PF
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.