Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Millicell™ EZ SlideHolder

Catalog No. PEZXMSH01 Shop All MilliporeSigma Products
Change view
Click to view available options
:
Millicell EZ Slide Microscope Slide Holder; Pk. of 1
Catalog No.
PEZXMSH01 Millicell EZ Slide Microscope Slide Holder; Pk. of 1
1 options

Catalog No. PEZXMSH01

Supplier: MilliporeSigma™ PEZXMSH01

Only null left
Add to Cart
Add to Cart

Culture, fix, stain, and view cells all in one device

  • Polypropylene wells are resistant to most fixatives—cells may be fixed and stained with wells still attached
  • Breakable tabs for easy well removal—no tools required to remove wells from slide
  • No adhesive used to attach wells to slide
  • No leaky wells
  • Raised corner ridges on lid facilitate stacking of multiple units in the incubator
  • Noncytotoxic and nonpyrogenic
  • Sterile

TRUSTED_SUSTAINABILITY

Specifications

Type EZ Slide
Color Black
Material Polystyrene
For Use With (Application) Millicell EZ slide microscope slide holder
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.