Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ HARLECO™ Gram Stain Reagents
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 Set
4 x 500 mL
500 mL
Type:
Crystal Violet
Decolorizer
Gram Iodine
Gram Stain Kit
Safranin
Description
- Use qualitative technique for differential staining of bacteria from cultures and patient specimens by modified Hucker Gram stain method
- Ideal for use with Midas III and other automated staining systems
- Gram Stain Set contains one 500mL bottle each of Crystal Violet, Safranin, Iodine, and Decolorizer Reagent
Order Info
Individual reagents supplied in packs of four 500mL bottles
Specifications
Specifications
| Type | Gram Stain Kit |
| Quantity | 1 Set |
| Includes | 500mL Crystal Violet, 500mL iodine, 500mL decolorizer, 500mL safranin |
| Format | Bottle |
| For Use With (Application) | Ideal for use with Midas III and other automated staining systems |
| Stain Type | Harleco |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction