Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ EZ-Fit™ Filtration Unit
Shop All MilliporeSigma Products
Click to view available options
Color:
Black with White Grid
White with Black Grid
Volume (Metric):
100 mL
250 mL
Individually wrapped:
Bulk Packed
Individually Wrapped
Description
- Designed for optimizing and securing laboratory workflow to provide time-saving and reliable microbiological results
- Comprise of 100mL or 250mL funnels
- Black or white gridded membranes
- After filtration of sample, membrane can be transferred and cultured on agar plate
- Sterilized and disposable
Conform to international standards (EP/USP) and comply with water testing regulations.
Specifications
Specifications
| Color | Black with White Grid |
| Material (Membrane) | MCE |
| Pore Size | 0.45 μm |
| Sterility | Sterile |
| Individually wrapped | Individually Wrapped |
| No. per Pack | 48 |
| Quantity | 48/Pk. |
| Type | Filtration Unit |
| Volume (Metric) | 250 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction