Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Enzyme Free Cell Dissociation Solution Hank's Based (1x)

Catalog No. S004C Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S004C 100 mL
S004B 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S004C Supplier MilliporeSigma™ Supplier No. S004C
Only null left
Add to Cart
Add to Cart

Hank's based formulation of chelating agents and agents to stabilize their activity on cells

Enzyme-Free Cell Dissociation Solution is a Hank's based formulation of chelating agents and agents to stabilize their activity on cells. This reagent does NOT contain porcine trypsin or any other protein. It works significantly faster than 0.5mM EDTA. In fact, it works as quickly as trypsin on most cells. Most importantly, it preserves the structural and functional integrity of cell surface proteins and does not have the cytotoxic effects sometimes associated with 0.5mm EDTA. It can be used to dissociate primary cells, tissues, and tumors with an increased plating efficiency.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Formulation Chelating and stabilizing agents
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.