Learn More
Biomatik Corporation Human FAM20C (Extracellular serine/threonine protein kinase FAM20C) ELISA Kit
Supplier: Biomatik Corporation EKF58388
Human FAM20C (Extracellular serine/threonine protein kinase FAM20C) ELISA Kit for detecting FAM20C in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8IXL6. Target Synonyms: Extracellular serine/threonine protein kinase FAM20C; Dentin matrix protein 4; DMP-4; Golgi casein kinase; Golgi-enriched fraction casein kinase; GEF-CK; DMP4; FAM20C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.