Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Non-esterified Free Fatty Acids (NEFA/FFA) Colorimetric Assay Kit

Catalog No. EEA017
Encompass_Preferred
Click to view available options
Quantity:
96 tests

Catalog No. EEA017

Supplier: Invitrogen™ EEA017

Only null left
Add to Cart
Add to Cart

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Non-esterified Free Fatty Acids (NEFA/FFA) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of NEFA in animal tissue samples. NEFAs are derived from adiopose tissue, where hormone-sensitive lipase decomposes triglycerides to product fatty acids. These acids circulate freely in the body and, when combined with plasma albumin, can be used as an energy sourced for muscles, organs, and the brain. They are also a substrate of fat synthesis, where the concentration of NEFAs is related to metabolism and endocrine function.

In this assay NEFA react with nantokite to form copper soap, which has a specific absorption peak at 715 nm. The content of NEFA can be calculated indirectly by measuring the OD value.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.15–1.5 mmol/L
Assay Sensitivity 0.15 mmol/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Tissue Homogenates
For Use With (Equipment) Microplate Reader
Incubation Time 3 min.
Interassay CV 5.1
Intraassay CV 3.3
Quantity 96 tests
Sample Volume 1000 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Non-esterified Free Fatty Acids (NEFA/FFA)
Total Assay Time 30 min.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.