Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ehrlich's Reagent, LabChem™
Click to view available options
Quantity:
1 L
Specifications
Specifications
| Chemical Name or Material | Ehrlich's Reagent |
| Color | Yellow |
| CAS | 71-23-8, 100-10-7 |
| CAS Max % | 95, 5 |
| Linear Formula | C9H11NO |
| UN Number | UN1993 |
| Solubility Information | Soluble in water |
| Formula Weight | 149.19 |
| Identification | Passes Test |
| Packaging | Amber Glass |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction