Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Radial Immunodiffusion Kit
SDPSCI_ED

Catalog No. s90248nd Shop All EDVOTEK Products
Click to view available options
Class Size:
10 Lab Groups

Catalog No. S90248ND

Supplier: Edvotek™ 273

Only null left
Add to Cart
Add to Cart

Includes

  • Antibody solution
  • Standard antigen solution
  • UltraSpec-Agarose™ powder and buffer powder
  • Unknown concentration of antigen
  • Sleeve of Petri dishes
  • Two 10mL pipets
  • 10 well cutters
  • 80 transfer pipets
  • 70 microtest tubes
  • Graph paper template
  • Practice loading solution

Requires

  • Automatic micropipets and tips (5 to 50μL)
  • Pipet pumps for 10mL pipets
  • Ruler
  • Plastic box or dish
  • Plastic wrap
  • Foil
  • Paper towels
  • Distilled water
  • Hotplate, Bunsen burner or microwave
  • 400 to 600mL beaker or Erlenmeyer flask
  • 150mL beaker or flask
  • Water bath (55°C [131°F])
  • 250mL graduated cylinder
  • 37°C (98.6°F) incubation oven

Introductory-level experiment quantitatively determines the unknown concentration of an antigen.

  • Antibody is incorporated into liquefied agar and allowed to gel; antigen is introduced by circular wells
  • Sensitive and quantitative

No experiment components contain human serum.

Specifications

Grade 9 to Undergraduate
For Use With (Application) To quantitatively determine the unknown concentration of an antigen
Includes Instructions, antigen and antibody, petri plates, pipets, well cutters, agarose, buffer, microtest tubes
Content And Storage Refrigerated; Store at: 4°C; 39°F
Class Size 10 Lab Groups
Product Type Radial Immunodiffusion Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.