Learn More
Research Products International Corp EDTA Disodium Salt [Ethylenediaminetetra-acetic acid, disodium salt dihydrate], 1 Kilogram

Supplier: Research Products International Corp E570201000.0
CAS Number: 6381-92-6
Molecular Formula: C10H14N2O8Na2 • 2H2O
Molecular Weight: 372.2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.