Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBV LMP1 Mouse anti-Virus, Unconjugated, Clone: S12, MilliporeSigma™
Mouse Monoclonal Antibody
$204.00 - $520.00
Specifications
| Antigen | EBV LMP1 |
|---|---|
| Clone | S12 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABF22482MI
|
MilliporeSigma
MABF224825UG |
25 μg |
Each for $204.00
|
|
|||||
|
MABF22481MI
|
MilliporeSigma
MABF2248100UG |
100 μg |
Each for $520.00
|
|
|||||
Description
EBV LMP1 Monoclonal antibody specifically detects EBV LMP1 in Virus samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting.Specifications
| EBV LMP1 | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Virus | |
| LMP1, BNLF1 | |
| IgG2a κ | |
| Protein G Purified | |
| Clone S12 is a mouse monoclonal antibody that specifically detects Latent membrane protein 1 (LMP1) in Epstein-Barr virus transformed cells. |
| S12 | |
| Monoclonal | |
| Purified | |
| Inflammation and Immunology | |
| Latent membrane protein;p63 | |
| A fusion protein (pKH548) consisting of 189 amino acids from the C-terminus region of LMP1 coupled to beta-galactosidase. | |
| Primary | |
| Stable for 1 year at 2 to 8°C from date of receipt. | |
| ∼59 kDa observed; 41.98 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title