Learn More
Invitrogen™ EB1/EB2/EB3 Monoclonal Antibody (KT65)

Description
The antibody detects a 30 kDa* protein corresponding to the molecular mass of EB1 on SDS-PAGE immunoblots of human HeLa, mouse brain, and rat PC12 cells. The antibody also detects EB1 by immunocytochemistry in HeLa and A431 cells. The antibody may weakly detect endogenous EB2 and EB3 in some cells and tissues.
Specifications
Specifications
| Antigen | EB1/EB2/EB3 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | KT65 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | Mapre1 |
| Gene Accession No. | Q15555, Q15691, Q3B8Q0, Q5XIT1, Q61166, Q66HR2, Q6PER3, Q8R001, Q9UPY8 |
| Gene Alias | APC; Microtubule RP/EB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.