Learn More
STEMCELL Technologies EasySep™ Human Memory B Cell Isolation Kit, For processing: 2 x 10^9 cells
Supplier: STEMCELL Technologies 17864
The EasySep™ Human Memory B Cell Isolation Kit is a two-step method designed to isolate CD27+ B cells from fresh or previously frozen peripheral blood mononuclear cells. First, CD27+ cells are isolated by column-free immunomagnetic positive selection using antibody complexes and EasySep™ Releasable RapidSpheres™. Then, bound magnetic particles are removed from the EasySep™-isolated CD27+ cells, and unwanted non-B cells are targeted for depletion using antibody complexes and EasySep™ Dextran RapidSpheres™. The final isolated fraction contains highly purified CD27+ memory B cells that are immediately ready for downstream applications. An optional protocol allows for the isolation of naïve B cells in parallel for use in functional studies.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.