Learn More
STEMCELL Technologies EasySep™ Human CD34 Positive Selection Kit II, For processing: 5 x 10^9 cells
Supplier: STEMCELL Technologies 17856RF
The EasySep™ Human CD34 Positive Selection Kit II is designed to isolate CD34+ cells from fresh or previously frozen bone marrow or mobilized peripheral blood mononuclear cells, or from previously frozen cord blood mononuclear cells, or from hESC/hiPSC-derived hematopoietic cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD34 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD34 antigen is expressed on hematopoietic stem and progenitor cells.This product can be used in place of the EasySep™ Human CD34 Positive Selection Kit (Catalog #18056) for even faster cell isolations.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.