Learn More
Boca Scientific Inc e-Myco PCR Mycoplasma Detection Kit, 48 Tubes
Supplier:  Boca Scientific Inc 25235
Encompass
The e-Myco Mycoplasma PCR Detection Kit consists of a set of primers designed to detect the presence of mycoplasma and determine the species. This kit can detect five common cell culture-infecting types of mycoplasma. The species of mycoplasma can be determined by sequencing the amplified products. 49 species can be detected. Each detection tube contains all components required for PCR in a dried and stable pellet format, including i-StarTaq DNA Polymerase, dNTPs, 10x Buffer, Mycoplasma primers, 8-methoxypsoralen, and an internal control for Mycoplasma partial gene amplifications. The reaction can begin by simply adding the template DNA or samples to be tested. PCR with this primer set is based on conserved 16S rRNA and delivers detection results in only a few hours. This is far more efficient than the conventional detection methods that involve culturing samples on selective media and require at least 1 week to obtain results.
Catalog No. 50-168-5641
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.