Learn More
New England Biolabs, Inc. E.coli DNA Ligase – 1000 units
SDP
Supplier:  New England Biolabs, Inc. M0205L
Encompass
E. coli DNA Ligase catalyzes the formation of a phosphodiester bond between the 5'-phosphate and the 3'-hydroxyl of two adjacent DNA strands in duplex DNA with cohesive ends. It is not appreciably active on blunt-ended substrates. E. coli DNA Ligase uses NAD as a cofactor and can be heat-inactivated.
Catalog No. 50-811-611
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.