Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ 400 Shirt
GREENER_CHOICE

Catalog No. 0136118E Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
01-361-18E 2X-Large
01-361-18A Small
01-361-18B Medium
01-361-18C Large
01-361-18D X-Large
19-042-277 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks, or in potentially flammable or explosive environments.
Catalog No. 01-361-18E Supplier DuPont™ Supplier No. TY303SWH2X005000
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks, or in potentially flammable or explosive environments.

Lightweight, chemically and biologically inert, tear-resistant

  • Shirt design with front snap closures for easy donning and doffing
  • Laydown collar
  • Open wrists
  • Typically worn with TY350S WH pants
  • Manufactured under specifications that do not contain natural rubber latex
  • D13397444,D13494332,D13494319,D13397433,D13397424,D13494249
TRUSTED_SUSTAINABILITY

Specifications

Size 2X-Large
Gender Unisex
Material Tyvek
Color White
Seam Type Serged
Closure Type Snaps
Product Type Shirt
For Use With (Application) Lead and Asbestos Abatement/Remedation, General Maintenance/Operations, Spray Painting, General Clean-up
Length (English) Sleeve 35.75 in.
Length (Metric) Sleeve 90.8 cm

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.