Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DTT, 1M

Catalog No. P2325
Encompass_Preferred
Click to view available options
Quantity:
1 mL

Catalog No. P2325

Supplier: Invitrogen™ P2325

Only null left
Add to Cart
Edge
Add to Cart

DTT, 1M

DTT is an essential component for kinase assays using LanthaScreen™ Technology. Invitrogen's DTT is 1M in water. It is tested for background fluorescence, making it optimal for use in high-throughput screening applications.
TRUSTED_SUSTAINABILITY

Specifications

Formulation 1M DTT in water
pH Range 6-8
Content And Storage DTT is shipped on dry ice and should be stored at -20°C.
Shipping Condition Dry Ice
Form Liquid
Quantity 1 mL
Product Type Dithiothreitol

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.