Learn More
Drummond™ Dialamatic Microdispensers
Catalog No. 2117015B
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
Increments:
1μL
2μL
10μL
Catalog No. 21-170-15B
Supplier: Drummond™ 3000225
Includes
100μL size comes with 50 glass bores; all others with 100 glass bores.
Permit accurate fractional deliveries
Four adjustable models cover volume range 1 to 100μL. Threaded plunger is graduated for accurate deliveries. Turn plunger to desired volume.
  • Two scales on 10 and 100μL sizes simplify volume selection
  • Accuracy to ±1% at full volume
  • Aluminum construction
  • Borosilicate glass bores with stainless steel plungers; PTFE tips on plungers 25μL or larger
Specifications
Accuracy ±1%
Material Aluminum
Number of Channels 1
Type Dialamatic Microdispenser
Volume (Metric) 25 μL
For Use With (Application) Adjustable, direct reading plunger sets volume
Includes 50 Borosilicate Glass bores with Stainless Steel plunger
Increments 1μL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.