Learn More
Pyramex™ DP3000 Series Ear Plugs
Catalog No. 19155296 Shop All Pyramex Safety Products Products
Click to view available options
:
Uncorded, Push-in
Catalog No. 19-155-296
Supplier: Pyramex™ DP3000
Push-in disposable ear plug promotes clean insertion even when wearer's hands are dirty
  • No ear plug roll down required
  • Stem eliminates foam contact with fingers
  • The soft foam ear plugs provide all day comfort
  • Contoured fit disposable polyurethane ear plugs
  • Packaged in a dispenser box
  • NRR 30 dB
Specifications
ANSI S3.1974 Yes
Size Universal
For Use With (Application) Assembly, Construction, Manufacturing, Transportation, Utilities, and Other Various Industries
High Visibility Yes
Packaging Type Dispenser box
Pairs per Package 50 Pairs/Pk.
Plug Material Foam
Plug Color Orange
Noise Reduction Rating (NRR) 30 dB
Corded Uncorded
Plug Shape Cone
Disposable Single-use
Metal Detectable No
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.