Learn More
Bellco Glass, Inc. The Dounce tissue homogenizer was designed for disruption of whole cells leaving cell nuclei undamaged. Two pestles are supplied with each complete unit. Large pestle is used for the initial sample reduction. The small pestle is used to form the final hom
Supplier: Bellco Glass, Inc. 198410002
The Dounce tissue homogenizer was designed for disruption of whole cells leaving cell nuclei undamaged. Two pestles are supplied with each complete unit. Large pestle is used for the initial sample reduction. The small pestle is used to form the final hom
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.