Learn More
United Scientific™ Double Buret Clamp
SDPSCI_ED
Catalog No. S90318 Shop All United Scientific Supplies, Inc Products
Click to view available options
Color:
Silver
Catalog No. S90318
Supplier: United Scientific™ CLBUR2N
Only null left
Add to Cart
Add to Cart
Double Buret Clamp with plastic-covered spring jaws protects glass from scratching.
  • Accepts burets up to 35 mm O.D.
  • Accepts support rods up to 0.5 in. O.D.
Specifications
Type Burette Clamp
Material Alloy Steel
Color Silver
Jaw Type Spring Loaded
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.