Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Isoborneol, 93%, tech.

Catalog No. AC122490250
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Catalog No. AC122490250

Supplier: Thermo Scientific Chemicals 122490250

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 124-76-5 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-76-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00074821
InChI Key DTGKSKDOIYIVQL-SZBHIRRCSA-N
Synonym dl-isoborneol
PubChem CID 126961757
IUPAC Name (1R,3R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
SMILES CC1(C2CCC1(C(C2)O)C)C

Specifications

Melting Point 208.0°C to 214.0°C
Color Yellow
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 91% min. (GC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 06, II, 80
Merck Index 15, 5173
Solubility Information Solubility in water: insoluble in water. Other solubilities: readily soluble in alcohol,chloroform and ether
Formula Weight 154.25
Percent Purity 93%
Grade Technical
Physical Form Powder
Chemical Name or Material DL-Isoborneol, tech., 93%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 204-712-4
RTECSNumber NP7300000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.