Learn More
Medchemexpress LLC HY-B0128 500mg Medchemexpress, Diphylline CAS:479-18-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0128500MG
Encompass_Preferred
Medchemexpress, HY-B0128 500mg Diphylline CAS:479-18-5 Diphylline (Diprophylline) is a potent A1/A2 adenosine receptor antagonist and cyclic nucleotide phosphodiesterase inhibitor. Diphylline, a xanthine derivative, is a bronchodilator and vasodilator drug and has the potential for chronic bronchitis and emphysema. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2373