Learn More
Medchemexpress LLC HY-10492 100mg Medchemexpress, Dinaciclib SCH 727965 CAS:779353-01-4 Purity:98%
Supplier: Medchemexpress LLC HY10492100MG
Medchemexpress,HY-10492 100mg Dinaciclib SCH 727965 CAS:779353-01-4 Formula:C21H28N6O2 Dinaciclib is a potent inhibitor of CDK, with IC50s of 1, 1, 3, and 4 nM for CDK2, CDK5, CDK1, and CDK9, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.