Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylglyoxime RS, 1% (w/v) Alcoholic Solution, Ricca Chemical
SDP

Catalog No. 25704 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle

Catalog No. 2570-4

Supplier: Ricca Chemical Company 25704

Only null left
Add to Cart
Add to Cart

For Nickel Determination

Chemical Identifiers

CAS 95-45-4, 64-17-5, 67-56-1
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002117
InChI Key JGUQDUKBUKFFRO-GGWOSOGESA-N
Synonym biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
PubChem CID 135399895
IUPAC Name (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine
SMILES C\C(=N/O)\C(\C)=N\O

Specifications

Color Colorless
CAS Min % 1.21, 97.0
CAS Max % 1.26, 100.0
Percent Purity 1%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Alcoholic Solution
Quantity 120 mL
Physical Form Liquid
Concentration or Composition Notes 1% (w/v)
Chemical Name or Material Dimethylglyoxime, Reagent Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.