Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Diclofenac sodium, 98%

Catalog No. AC445250100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC445250100 10 g Glass bottle
1 options

Catalog No. AC445250100

Supplier: Thermo Scientific Chemicals 445250100

Only null left
Add to Cart

Diclofenac sodium, CAS # 15307-79-6, is a nonsteroidal anti-inflammatory drug (NSAID) that inhibits prostaglandin synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Diclofenac is a is a nonsteroidal anti-inflammatory drug (NSAID) with anti-inflammatory and analgesic properties
  • Diclofenac has been shown to have anti-tumor capabilities through inhibition of MYC expression and glucose metabolism in models of melanoma and leukemia

Applications

  • Diclofenac sodium can be used as an antioxidant and free radical scavenger
  • It has been shown to have anti-bacterial activity against Listeria, E. coli, and M. tuberculosis partially through inhibition of bacterial DNA synthesis
  • It can be used for the study of metabolic activity as a CYP2C9 substrate
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15307-79-6
Molecular Formula C14H10Cl2NNaO2
Molecular Weight (g/mol) 318.13
InChI Key KPHWPUGNDIVLNH-UHFFFAOYSA-M
Synonym diclofenac sodium, sodium diclofenac, diclofenac sodium salt, voltaren, solaraze, anthraxiton, prophenatin, allvoran, batafil, delimon
PubChem CID 5018304
ChEBI CHEBI:4509
IUPAC Name sodium;2-[2-(2,6-dichloroanilino)phenyl]acetate
SMILES C1=CC=C(C(=C1)CC(=O)[O-])NC2=C(C=CC=C2Cl)Cl.[Na+]

Specifications

Melting Point 283.0°C to 285.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Merck Index 15,3091
Solubility Information Solubility in water: 50mg/mL
Formula Weight 318.13
Percent Purity 98%
Chemical Name or Material Diclofenac sodium
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 239-346-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.