Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dickson™ Chart Recorder Red Replacement Pen
SDP

Catalog No. 13940997
Click to view available options
Color:
Red

Catalog No. 13-940-997

Supplier: Dickson™ P222

Only null left
Add to Cart
Add to Cart

For Use With: Large capacity refrigerated Incubator

  • Accessory for use with Dickson Automatic Chart Recorders

Specifications

Color Red
For Use With (Equipment) Large capacity refrigerated incubator
Type Chart Recorder Pen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.