Learn More
Diamedix™ Immunosimplicity™ Measles IgG Test Kit
SDP
Catalog No. 23451036
Encompass
Click to view available options
Quantity:
96 Tests
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Catalog No. 23-451-036
Supplier: Diamedix™ 720520
Only null left
Add to Cart
Add to Cart
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes
Antigen Wells, Cut-off Calibrator, Positive Control, Negative Control, Sample A Diluent, Wash S Concentrate (20X), Conjugate, Substrate HRP, Stop M Solution
Detects IgG antibodies to measles virus in human serum
The Diamedix™ Immunosimplicity™ Measles IgG Test Kit is a semi-quantitative indirect enzyme immunoassay
  • Detects IgG antibodies to measles virus in human serum
  • Aids in the assessment of the patient's immunological response to measles virus
  • Also aids in the qualitative determination of the immune status of individuals and, when evaluating paired sera, as an aid in the diagnosis of measles infection
  • For in vitro diagnostic use
  • Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
Detectable Analytes Measles virus IgG
Type Immunosimplicity™ Measles IgG Test Kit
Includes Microwell 12 x 8-well strips, positive control 1.8mL/vial, cut-off calibrator 1.8mL/ vial, negative control 1.8mL/vial, conjugate 13mL, Wash Buffer 50mL, substrate 13mL, stop solution 13mL
Quantity 96 Tests
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.