Learn More
Aviva Systems Biology Corp DHX16 Antibody - N-terminal region (ARP37711_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37711_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHX16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DHX16. Alternate names: DBP2, PRP8, Prp2, DDX16, PRPF2, PRO2014ApplicationWB. Species reactivity: Cow, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56402