Learn More
Foxx Life Sciences Borosil Glass Desiccator with Porcelain Plate and Borosilicate Lid with Plastic Knob, Large (L), 300mm Diameter, 1/EA
Supplier: Foxx Life Sciences 3082045
From streamlined contours to interchangeable lids and bodies, Borosil Non-Vacuum Desiccators reflect great care and attention to detail. Supplied with a porcelain plate and ground glass flanges, these desiccators are perfect for drying, and for storage of moisture-sensitive substances. Compatible with any common desiccant. Avail. in inner diameters from 100mm (XS) to 300mm (XL). Borosil Desiccators are also Avail. in Light-Blocking Amber. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free and incredibly resistant to thermal shock.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.