Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Decon™ Phenazine pH Test Paper
Shop All Decon Labs Products

Click to view available options
pH Range:
4 to 8
Description
- Phenazine paper is designed for determining pH values
- Provides sharp, clear changes within pH range of 4.5 to 7.5
- Color scale chart in increments of 0.5pH units
Specifications
Specifications
| pH Range | 4 to 8 |
| For Use With (Application) | Designed for determining pH values, provides sharp, clear changes within pH range of 4 to 8, for in-vitro diagnostic use |
| Includes | Comparative color scale chart in increments of 0.5 pH units |
| Quantity | 1 Roll |
| Type | Phenazine pH Indicator Paper |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction